Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD299 (DC-SIGN/L) Recombinant Mouse Monoclonal Antibody (16E7)
GREENER_CHOICE

Catalog No. PIMA547779
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547779 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547779 Supplier Invitrogen™ Supplier No. MA547779
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

Specificity: Human CD299, which is an oligomeric type II transmembrane protein with a C-type lectin extracellular domain, the expression of which is restricted to immature DC, macrophages in the lung, and endothelial cells in the liver. It binds ICAM-3 and ICAM-7 to mediate the interaction of DC with T lymphocytes and endothelial cells in the initial stages of immune response and in the migratory behavior of DC. CD299 also binds the gp120 protein of HIV and the E2 envelope protein of HCV, thereby playing a role in viral infection.

CLEC4M is a type II integral membrane protein that is 77% identical to CD209 antigen, a HIV gp120-binding protein. This protein, like CD209, efficiently binds both intercellular adhesion molecule 3 (ICAM3) and HIV-1 gp120, and enhances HIV-1 infection of T cells.Western blots using two different antibodies against two unique regions of this protein target confirm the same apparent molecular weight in our tests.This gene encodes a type II integral membrane protein that is 77% identical to CD209 antigen, a HIV gp120-binding protein. This protein, like CD209, efficiently binds both intercellular adhesion molecule 3 (ICAM3) and HIV-1 gp120, and enhances HIV-1 infection of T cells. This gene is mapped to 19p13.3, in a cluster with the CD209 and CD23/FCER2 genes. Multiple alternatively spliced transcript variants have been found for this gene, but the biological validity of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD299 (DC-SIGN/L)
Applications Flow Cytometry, Immunohistochemistry
Classification Recombinant Monoclonal
Clone 16E7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene CLEC4M
Gene Accession No. Q9H2X3
Gene Alias CD209 antigen-like protein 1; Cd209b; CD209b antigen; CD209L; CD209L1; CD299; CD299 antigen; CLEC4M; C-type lectin domain family 4 member M; C-type lectin domain family 4, member M; DC-SIGN2; DCSIGNR; DC-SIGNR; DC-SIGN-related protein; Dendritic cell-specific ICAM-3-grabbing non-integrin 2; HP10347; liver/lymph node-specific ICAM-3 grabbing non-integrin; liver/lymph node-specific ICAM-3-grabbing non-integrin; LSIGN; L-SIGN; mannose binding C-type lectin DC-SIGNR; MGC129964; MGC47866
Gene Symbols CLEC4M
Host Species Mouse
Immunogen CD299.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10332
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.