Learn More
Invitrogen™ CD299 (DC-SIGN/L) Recombinant Rabbit Monoclonal Antibody (6K3W9)
Rabbit Recombinant Monoclonal Antibody
$169.00 - $419.00
Specifications
Antigen | CD299 (DC-SIGN/L) |
---|---|
Clone | 6K3W9 |
Concentration | 1.26 mg/mL |
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | ELISA, Western Blot |
Description
Immunogen sequence: QVSKVPSSLS QEQSEQDAIY QNLTQLKAAV GELSEKSKLQ EIYQELTQLK AAVGELPEKS KLQEIYQELT RLKAAVGELP EKSKLQEIYQ ELTRLKAAVG ELPEKSKLQE IYQELTRLKA AVGELPEKSK LQEIYQELTE LKAAVGELPE KSKLQEIYQE LTQLKAAVGE LPDQSKQQQI YQELTDLKTA FERLCRHCPK DWTFFQGNCY FMSNSQRNWH DSVTACQEVR AQLVVIKTAE EQNFLQLQTS RSNRF.
CLEC4M is a type II integral membrane protein that is 77% identical to CD209 antigen, a HIV gp120-binding protein. This protein, like CD209, efficiently binds both intercellular adhesion molecule 3 (ICAM3) and HIV-1 gp120, and enhances HIV-1 infection of T cells.Western blots using two different antibodies against two unique regions of this protein target confirm the same apparent molecular weight in our tests.This gene encodes a type II integral membrane protein that is 77% identical to CD209 antigen, a HIV gp120-binding protein. This protein, like CD209, efficiently binds both intercellular adhesion molecule 3 (ICAM3) and HIV-1 gp120, and enhances HIV-1 infection of T cells. This gene is mapped to 19p13.3, in a cluster with the CD209 and CD23/FCER2 genes. Multiple alternatively spliced transcript variants have been found for this gene, but the biological validity of some variants has not been determined.Specifications
CD299 (DC-SIGN/L) | |
1.26 mg/mL | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
Q9H2X3 | |
10332 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 71-399 of human DC-SIGNR/CD299 (NP_055072.3). | |
Antibody |
6K3W9 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human | |
CLEC4M | |
CD209 antigen-like protein 1; Cd209b; CD209b antigen; CD209L; CD209L1; CD299; CD299 antigen; CLEC4M; C-type lectin domain family 4 member M; C-type lectin domain family 4, member M; DC-SIGN2; DCSIGNR; DC-SIGNR; DC-SIGN-related protein; Dendritic cell-specific ICAM-3-grabbing non-integrin 2; HP10347; liver/lymph node-specific ICAM-3 grabbing non-integrin; liver/lymph node-specific ICAM-3-grabbing non-integrin; LSIGN; L-SIGN; mannose binding C-type lectin DC-SIGNR; MGC129964; MGC47866 | |
CLEC4M | |
Primary | |
Protein A |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.