Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD2AP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596807
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596807 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596807 Supplier Invitrogen™ Supplier No. PA596807
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-431, Raji, MCF7, SW480, HL-60, Mouse kidney, Rat kidney; Cellular Location: Cell projection, Cytoplasm, cytoskeleton, ruffle Immunogen sequence: KYFSLKPEEK DEKSTLEQKP SKPAAPQVPP KKPTPPTKAS NLLRSSGTVY PKRPEKPVPP PPPIAKINGE VSSISSKFET EPVSKLKLDS EQLPLRPKSV DFDSLTVRTS KETDVVNFDD IASSENLLHL TANRPKMPGR RLPGRFNGGH SPTHSPEKIL KLPKEEDSAN LKPSELKKDT CYSPKPSVYL STPSSASKAN TTAFLTPLEI KAKVETDDVK KNSLDELRAQ IIELLCIVEA LKKDHGKELE KLRKDLEEEK TMRSNLEMEI EKLKKAVLSS.

This gene encodes a scaffolding molecule that regulates the actin cytoskeleton. The protein directly interacts with filamentous actin and a variety of cell membrane proteins through multiple actin binding sites, SH3 domains, and a proline-rich region containing binding sites for SH3 domains. The cytoplasmic protein localizes to membrane ruffles, lipid rafts, and the leading edges of cells. It is implicated in dynamic actin remodeling and membrane trafficking that occurs during receptor endocytosis and cytokinesis. Haploinsufficiency of this gene is implicated in susceptibility to glomerular disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD2AP
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CD2AP
Gene Accession No. F1LRS8, Q9JLQ0, Q9Y5K6
Gene Alias adapter protein CMS; AL024079; C78928; Cas ligand with multiple SH3 domains; Cas ligand with multiple Src homology 3 (SH3) domains; CD2 associated protein; Cd2ap; CD2-associated protein; CMS; mesenchyme to epithelium transition protein, SH3 domains; mesenchyme-to-epithelium transition protein with SH3 domains 1; Mets1; METS-1; SH3 domain-containing adapter protein; SH3 domain-containing adaptor protein METS-1
Gene Symbols CD2AP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 360-639 of human CD2AP (NP_0362521).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12488, 23607, 316258
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.