Learn More
Invitrogen™ CD2AP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578975
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Jurkat whole cell, human A431 whole cell, human Hela whole cell, human K562 whole cell. IHC: mouse kidney tissue, rat kidney tissue. ICC/IF: Hela cell IP: Jurkat cell. Flow: K56 cell.
This gene encodes a scaffolding molecule that regulates the actin cytoskeleton. The protein directly interacts with filamentous actin and a variety of cell membrane proteins through multiple actin binding sites, SH3 domains, and a proline-rich region containing binding sites for SH3 domains. The cytoplasmic protein localizes to membrane ruffles, lipid rafts, and the leading edges of cells. It is implicated in dynamic actin remodeling and membrane trafficking that occurs during receptor endocytosis and cytokinesis. Haploinsufficiency of this gene is implicated in susceptibility to glomerular disease.
Specifications
CD2AP | |
Polyclonal | |
Unconjugated | |
CD2AP | |
adapter protein CMS; AL024079; C78928; Cas ligand with multiple SH3 domains; Cas ligand with multiple Src homology 3 (SH3) domains; CD2 associated protein; Cd2ap; CD2-associated protein; CMS; mesenchyme to epithelium transition protein, SH3 domains; mesenchyme-to-epithelium transition protein with SH3 domains 1; Mets1; METS-1; SH3 domain-containing adapter protein; SH3 domain-containing adaptor protein METS-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12488, 23607, 316258 | |
-20°C | |
Lyophilized |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
500 μg/mL | |
PBS with 4mg trehalose and no preservative | |
F1LRS8, Q9JLQ0, Q9Y5K6 | |
CD2AP | |
E. coli-derived human CD2AP recombinant protein (Position: K253-K337). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.