Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD2APRabbit anti-Human, Polyclonal, Rockland™
SDP

Catalog No. 501992280
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2280 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-2280 Supplier Rockland Immunochemicals Supplier No. 600401J96
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. A BLAST analysis was used to suggest cross-reactivity with human based on 100% sequence homology. Cross-reactivity with CD2AP from other sources has not been determined.

This gene encodes a scaffolding molecule that regulates the actin cytoskeleton. The protein directly interacts with filamentous actin and a variety of cell membrane proteins through multiple actin binding sites, SH3 domains, and a proline-rich region containing binding sites for SH3 domains. The cytoplasmic protein localizes to membrane ruffles, lipid rafts, and the leading edges of cells. It is implicated in dynamic actin remodeling and membrane trafficking that occurs during receptor endocytosis and cytokinesis. Haploinsufficiency of this gene is implicated in susceptibility to glomerular disease.

Specifications

Antigen CD2AP
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl, 1mg/mL BSA, 50% glycerol and 0.01% sodium azide; pH 7.2
Gene CD2AP
Gene Accession No. Q9Y5K6
Gene Alias Adapter protein CMS; AL024079; C78928; Cas ligand with multiple SH3 domains; Cas ligand with multiple Src homology 3 (SH3) domains; CD2 associated protein; Cd2ap; CD2-associated protein; CMS; mesenchyme to epithelium transition protein, SH3 domains; Mesenchyme-to-epithelium transition protein with SH3 domains 1; Mets1; METS-1; SH3 domain-containing adapter protein; SH3 domain-containing adaptor protein METS-1
Gene Symbols CD2AP
Host Species Rabbit
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23607
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.