Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD2BP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563947
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563947 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563947 Supplier Invitrogen™ Supplier No. PA563947
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPKRKVTFQG VGDEEDEDEI IVPKKKLVDP VAGSGGPGSR FKGKHSLDSD EE Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 94%.

This gene encodes a bi-functional protein. In the cytoplasm, the encoded protein binds the cytoplasmic tail of human surface antigen CD2 via its C-terminal GYF domain, and regulate CD2-triggered T lymphocyte activation. In the nucleus, this protein is a component of the U5 small nuclear ribonucleoprotein complex and is involved in RNA splicing. A pseudogene has been identified on chromosome 7. Alternative splicing results in multiple transcript variants but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD2BP2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CD2BP2
Gene Accession No. O95400
Gene Alias 1500011B02Rik; 2410024K20Rik; AW048183; CD2 (cytoplasmic tail) binding protein 2; CD2 antigen (cytoplasmic tail) binding protein 2; CD2 antigen cytoplasmic tail-binding protein 2; CD2 cytoplasmic domain-binding protein 2; CD2 cytoplasmic tail binding protein 2; CD2 tail-binding protein 2; CD2BP2; FWP010; KIAA1178; LIN1; PPP1R59; protein phosphatase 1, regulatory subunit 59; Snu40; U5 snRNP 52K protein; U5-52K
Gene Symbols CD2BP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CD2BP2. Recombinant protein control fragment (Product #RP-103864).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10421
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.