Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD2BP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518286
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518286 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518286 Supplier Invitrogen™ Supplier No. PA518286
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

This gene encodes a bi-functional protein. In the cytoplasm, the encoded protein binds the cytoplasmic tail of human surface antigen CD2 via its C-terminal GYF domain, and regulate CD2-triggered T lymphocyte activation. In the nucleus, this protein is a component of the U5 small nuclear ribonucleoprotein complex and is involved in RNA splicing. A pseudogene has been identified on chromosome 7. Alternative splicing results in multiple transcript variants but their biological validity has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD2BP2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene CD2BP2
Gene Accession No. O95400
Gene Alias 1500011B02Rik; 2410024K20Rik; AW048183; CD2 (cytoplasmic tail) binding protein 2; CD2 antigen (cytoplasmic tail) binding protein 2; CD2 antigen cytoplasmic tail-binding protein 2; CD2 cytoplasmic domain-binding protein 2; CD2 cytoplasmic tail binding protein 2; CD2 tail-binding protein 2; CD2BP2; FWP010; KIAA1178; LIN1; PPP1R59; protein phosphatase 1, regulatory subunit 59; Snu40; U5 snRNP 52K protein; U5-52K
Gene Symbols CD2BP2
Host Species Goat
Immunogen Synthetic peptide sequence (PKRKVTFQGVGDE) corresponding to the N-terminus amino acids of CD2BP2 (aa 2-15).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10421
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.