Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 Antibody (CA17.2A12), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP249891AF700
Description
CD3 Monoclonal specifically detects CD3 in Canine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence.Specifications
CD3 | |
Monoclonal | |
Alexa Fluor 700 | |
PBS with 0.05% Sodium Azide | |
CD3E | |
Affinity enriched TCR/CD3 membrane proteins isolated from thymocytes and the T cell line CLGL-90 | |
0.1 mL | |
Primary | |
Canine | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence | |
CA17.2A12 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence | |
CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, CD3G, CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ18683, FLJ79544, FLJ94613, MGC138597, T3DOKT3, delta chain, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
Mouse | |
Protein A or G purified | |
Adaptive Immunity, Apoptosis, Autoimmune Diseases, Diabetes Research, Immunology, Inflammation, Innate Immunity, Signal Transduction, Stem Cells | |
916 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction