Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 Antibody (KT3), HRP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10064871H
Description
CD3 Monoclonal specifically detects CD3 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD3 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
CD3E | |
CBAT6 thymocytes | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
IgG2a |
Immunohistochemistry, Immunohistochemistry (Frozen) | |
KT3 | |
Immunohistochemistry, Immunohistochemistry-Frozen | |
CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, CD3G, CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ18683, FLJ79544, FLJ94613, MGC138597, T3DOKT3, delta chain, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
Rat | |
21.4 kDa | |
0.125 mL | |
Adaptive Immunity, Apoptosis, Autoimmune Diseases, Diabetes Research, Immunology, Inflammation, Innate Immunity, Signal Transduction, Stem Cells | |
The antibody recognizes mouse CD3 antigen. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction