Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 Antibody (OKT3) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 9 publications

$192.47 - $371.72

Specifications

Antigen CD3
Clone OKT3
Concentration 1.0 mg/ml
Dilution Western Blot reported in scientific literature (PMID 28699640), Flow Cytometry, Functional, Immunoblotting reported in scientific literature (PMID 23880762), In vitro assay, In vivo assay
Applications Western Blot, Flow Cytometry, Functional Assay, Immunoblot, In vitro Assay
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP225186SS
SDP
View Documents
Novus Biologicals
NBP225186SS
0.025 mg
Each for $192.47
Only null left
Add to Cart
 
NBP225186
SDP
View Documents
Novus Biologicals
NBP225186
0.1 mg
Each for $371.72
Only null left
Add to Cart
 
Description

Description

CD3 Monoclonal specifically detects CD3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Functional, Immunoblotting, In vitro assay, In vivo assay.
Specifications

Specifications

CD3
1.0 mg/ml
Western Blot, Flow Cytometry, Functional Assay, Immunoblot, In vitro Assay
Unconjugated
Mouse
Adaptive Immunity, Apoptosis, Autoimmune Diseases, Immunology, Inflammation, Innate Immunity, Signal Transduction, Stem Cells
NP_000723
916
Human PBMCs were used as immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
23.1 kDa
OKT3
Western Blot reported in scientific literature (PMID 28699640), Flow Cytometry, Functional, Immunoblotting reported in scientific literature (PMID 23880762), In vitro assay, In vivo assay
Monoclonal
Purified
RUO
Human, Mouse
CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, CD3G, CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ18683, FLJ79544, FLJ94613, MGC138597, T3DOKT3, delta chain, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
CD3
IgG2a κ
Protein G purified
This product reacts with Human.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.