Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 Antibody (OKT3) - Low Endotoxin, Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-4964565 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224867SS 0.025 mg
NBP224867 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224867SS Supplier Novus Biologicals Supplier No. NBP224867SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 8 publications

CD3 Monoclonal specifically detects CD3 in Human, Mouse samples. It is validated for CyTOF, Flow Cytometry, Flow Cytometry (Cell Surface), Functional Assay, Immunoblot, In vitro Assay, In vivo Assay.

Specifications

Antigen CD3
Applications CyTOF, Flow Cytometry, Flow Cytometry (Cell Surface), Functional Assay, Immunoblot, In vitro Assay, In vivo Assay
Classification Monoclonal
Clone OKT3
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1 uL/ 1 million cells, Functional, Immunoblotting reported in scientific literature (PMID 23880762), In vitro assay reported in scientific literature (PMID 26084721), In vivo assay reported in scientific literature (PMID 25535359), Flow (Cell Surface), CyTOF-ready
Gene Accession No. P04234
Gene Alias anti-CD3 okt3, CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, CD3G, CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ18683, FLJ79544, FLJ94613, MGC138597, okt3, okt3 ab, okt3 CD3, okt3 hybridoma, T3DOKT3, delta chain, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Gene Symbols CD3
Host Species Mouse
Immunogen Human PBMCs were used as immunogen.
Molecular Weight of Antigen 23.1 kDa
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Autoimmune Diseases, Immunology, Inflammation, Innate Immunity, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 916
Test Specificity Functional activation was utilized in the following publication: An Epstein-Barr Virus (EBV) mutant with enhanced BZLF1 expression causes lymphomas with abortive lytic EBV infection in a humanized mouse model. Ma D S, X Yu, J E Mertz, J E Gumperz, E Reinheim, Y Zhou, W Tang, W J Burlingham, M L Gulley, S C Kenney. J Virol 86 7976-7987 (2012). This antibody is reactive to Human.
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.