Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 Antibody (RM0027-3B19), Alexa Fluor™ 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212159AF350
Description
CD3 Monoclonal specifically detects CD3 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| CD3 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CD3E | |
| Mouse CD3 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RM0027-3B19 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, CD3G, CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ18683, FLJ79544, FLJ94613, MGC138597, T3DOKT3, delta chain, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| Rat | |
| 21.4 kDa | |
| Adaptive Immunity, Apoptosis, Autoimmune Diseases, Diabetes Research, Immunology, Inflammation, Innate Immunity, Signal Transduction, Stem Cells | |
| 916 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction