Learn More
Invitrogen™ CD3/CD44 Antibody Cocktail, FITC, PE
Description
This antibody is a monoclonal mixture of FITC-conjugated CD3 (clone M2AB, isotype IgG1) and PE-conjugated CD44 (clone 7F1, isotype IgG1).
Specifications
Specifications
| Antigen | CD3/CD44 |
| Applications | Flow Cytometry |
| Classification | Cocktail |
| Clone | M2AB, 7F1, 7F1 |
| Conjugate | FITC/PE |
| Formulation | PBS with 0.2% carrier protein and 0.08% sodium azide |
| Gene | CD3G |
| Gene Accession No. | P04234, P07766, P09693, P16070 |
| Gene Alias | CD3 antigen, delta subunit; CD3 delta; CD3 epsilon; CD3D; CD3d antigen, delta polypeptide (TiT3 complex); CD3d molecule; CD3d molecule, delta (CD3-TCR complex); CD3-DELTA; CD3E; CD3e antigen, epsilon polypeptide (TiT3 complex); CD3e molecule; CD3e molecule, epsilon (CD3-TCR complex); CD3-epsilon; CD3g; CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule; CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); CD3-GAMMA; CD44; CD44 antigen; CD44 molecule; CD44 molecule (Indian blood group); CDw44; cell surface glycoprotein CD44; chondroitin sulfate proteoglycan 8; CSPG8; ECMRIII; ECMR-III; Epican; extracellular matrix receptor III; FLJ18683; GP90 lymphocyte homing/adhesion receptor; HCAM; HCELL; hematopoietic cell E- and L-selectin ligand; heparan sulfate proteoglycan; Hermes antigen; homing function and Indian blood group system; HUTCH-I; hyaluronan receptor; Hyaluronate receptor; IMD17; IMD18; IMD19; IN; LHR; MC56; MDU2; MDU3; MIC4; OKT3, delta chain; PGP I; Pgp1; PGP-1; PGPI; PGP-I; Phagocytic glycoprotein 1; Phagocytic glycoprotein I; sCD 44; sCD44; soluble CD44; T3D; T3E; T3G; T-cell antigen receptor complex, epsilon subunit of T3; T-cell antigen receptor complex, gamma subunit of T3; T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; T-cell surface antigen T3/Leu-4 epsilon chain; T-cell surface glycoprotein CD3 delta chain; T-cell surface glycoprotein CD3 epsilon chain; T-cell surface glycoprotein CD3 gamma chain; TCRE |
| Gene Symbols | CD3D, CD3E, CD3G, CD44 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.