Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 delta Antibody (138), Alexa Fluor™ 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289730AF594
Description
CD3 delta Monoclonal specifically detects CD3 delta in Human samples. It is validated for Western Blot.Specifications
CD3 delta | |
Monoclonal | |
Alexa Fluor 594 | |
CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, T3DOKT3, delta chain, T-cell receptor T3 delta chain, T-cell surface glycoprotein CD3 delta chain | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD3 delta (NP_000723.1; Met1-Ala105). | |
0.1 mL | |
Adaptive Immunity, Apoptosis, Diabetes Research, Immunology, Innate Immunity, Signal Transduction | |
915 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
138 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction