Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 delta Antibody (1F4), Novus Biologicals™
SDP

Catalog No. NB10065230 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065230 0.1 mg
NB017697 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10065230 Supplier Novus Biologicals Supplier No. NB10065230
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD3 delta Monoclonal specifically detects CD3 delta in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CD3 delta
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 1F4
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:25, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10, Immunohistochemistry-Frozen 1:10-1:25
Gene Alias CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, T3DOKT3, delta chain, T-cell receptor T3 delta chain, T-cell surface glycoprotein CD3 delta chain
Gene Symbols CD3D
Host Species Mouse
Immunogen F344 rat T cells stimulated with PMA (TPA) and calcium ionophore
Purification Method IgM purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Diabetes Research, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 915
Test Specificity Recognizes Rat CD3D (Gene ID: 25710, UniProt: P19377) and Rat CD3G (Gene ID: 300678, UniProt: Q64159) NB100-65230 recognizes rat CD3, a 25kD antigen which is found on rat T-cells. NB100-65230 does not react with rat B cells. In immunohistology it stains rat thymus tissues strongly in the medulla and weakly in the cortex. Functionally the addition of the antibody to a culture of rat T cells induces the proliferation of T-cells in the presence of PMA.
Target Species Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.