Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 delta Antibody (6), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289729MFV450
Description
CD3 delta Monoclonal antibody specifically detects CD3 delta in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
CD3 delta | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunoprecipitation | |
6 | |
Western Blot, ELISA, Immunoprecipitation | |
CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, T3DOKT3, delta chain, T-cell receptor T3 delta chain, T-cell surface glycoprotein CD3 delta chain | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD3 delta (Accession#: NP_000723.1; Met 1-Ala 105). | |
0.1 mL | |
Adaptive Immunity, Apoptosis, Diabetes Research, Immunology, Innate Immunity, Signal Transduction | |
915 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction