Learn More
CD3ε (Early T-Cell Marker) Ab-1 Rabbit Polyclonal Antibody, Epredia™
Obtain accurate and reproducible results in immunohistochemistry applications with Epredia™ CD3ε (Early T-Cell Marker) Ab-1, Rabbit Polyclonal Antibody.
Specifications
| Antigen | CD3e |
|---|---|
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
|
RB360A0
|
Epredia
RB360A0 |
Concentrated | 100 μL | N/A | N/A | N/A | |||
|
RB360A1
|
Epredia
RB360A1 |
Concentrated | 500 μL | N/A | N/A | N/A | |||
|
RB360R1
|
Epredia
RB360R1 |
Ready-to-Use | 1 mL | N/A | N/A | N/A | |||
|
RB360R7
|
Epredia
RB360R7 |
Ready-to-Use | 7 mL | N/A | N/A | N/A | |||
Description
CD3 consists of five different polypeptide chains (designated as γ, δ, ε, ζ, and η). The CD3 complex is closely associated at the lymphocyte cell surface with the T cell antigen receptor (TCR). It is involved in signal transduction to the T cell interior following antigen recognition. CD3 antigen is a highly specific marker for T cells, and is present in majority of T cell neoplasms.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Does not react with rat. Others not known.
Epitope: aa 156-168 of ε-chain
Immunogen: A synthetic 13-mer peptide, corresponding to aa 156-168 of the ε-chain of human CD3 protein
Molecular Weight: âˆ1⁄220kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| CD3e | |
| Polyclonal | |
| Rabbit | |
| T-Cell | |
| A synthetic 13-mer peptide, corresponding to aa 156-168 of the epsilon-chain of human CD3 protein | |
| Primary |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Human | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.