Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD3ε (Early T-Cell Marker) Ab-2, Mouse Monoclonal Antibody

Catalog No. MS401S0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
Form:
Concentrated
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS401S0 100 μL Concentrated
MS401S1 500 μL Concentrated
MS401S 1 mL Concentrated
MS401R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS401S0 Supplier Epredia™ Supplier No. MS401S0

Obtain accurate and reproducible results in immunohistochemistry experiments with Epredia™ CD3ε (Early T-Cell Marker) Ab-2, Mouse Monoclonal Antibody.

CD3 consists of five different polypeptide chains (designated as γ, δ, ε, ζ, and η). The CD3 complex is closely associated at the lymphocyte cell surface with the T cell antigen receptor (TCR). It is involved in signal transduction to the T cell interior following antigen recognition. CD3 antigen is a highly specific marker for T cells, and is present in the majority of T cell neoplasms.

Host Species: Mouse

Clone: PS1

Isotype: IgG2a

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: ε-chain

Immunogen: Recombinant nonglycosylated ε-chain of CD3 protein

Molecular Weight: ∽20 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD3e
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PS1
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant nonglycosylated epsilon-chain of CD3 protein
Quantity 100 μL
Regulatory Status IVD
Research Discipline T-Cell
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.