Learn More
Epredia™ Lab Vision™ CD3ε (Early T-Cell Marker) Ab-9, Mouse Monoclonal Antibody
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: F7.2.38
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Cytoplasmic region of ε-chain
Immunogen: Purified CD3 ε γ δ/CD3ω
Molecular Weight: ∽20kDa
Positive Control: Tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | CD3e |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | F7.2.38 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Purified CD3 epsilon gamma delta/CD3omega |
| Purification Method | Tonsil |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.