Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 epsilon Antibody (145-2C11), Alexa Fluor™ 532, Novus Biologicals™

Hamster Monoclonal Antibody
Supplier: Novus Biologicals FAB484X
Description
CD3 epsilon Monoclonal antibody specifically detects CD3 epsilon in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Cell depletion, T Cell Stimulation, CyTOF-reportedSpecifications
CD3 epsilon | |
Monoclonal | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Hamster | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Immunoprecipitation, Depletion Assay, T-Cell Activation, CyTOF | |
145-2C11 | |
Flow Cytometry, Immunoprecipitation, Cell depletion, T Cell Stimulation, CyTOF-reported | |
CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
H-2Kb-specific mouse cytotoxic T-lymphocyte | |
0.1 mL | |
Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
916 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction