Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 epsilon Antibody (145-2C11) - Chimeric - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. p-7106009 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25264125 0.025 mg
NBP25264102 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25264125 Supplier Novus Biologicals Supplier No. NBP2526410.025MG
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

CD3 epsilon Monoclonal specifically detects CD3 epsilon in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Functional, CyTOF-ready.

Specifications

Antigen CD3 epsilon
Applications Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 145-2C11
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vivo assay, CyTOF-ready
Formulation PBS with 0.02% Proclin 300. with 0.02% Proclin 300
Gene Alias CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Gene Symbols CD3E
Host Species Rabbit
Immunogen H-2Kb-specific murine cytotoxic T-lymphocyte (CTL) clone.
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Lines, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 916
Test Specificity Recognises a 25-kDa protein component (CD3e, originally called T3-e) of the antigen-specific T-cell receptor.
Target Species Mouse
Content And Storage Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.