Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 epsilon Antibody (500A2) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
| Antigen | CD3 epsilon |
|---|---|
| Clone | 500A2 |
| Dilution | Flow Cytometry 1:10 - 1:1000, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25264325
![]() |
Novus Biologicals
NBP2526430.025MG |
0.025 mg |
Each for $268.00
|
|
|||||
NBP25264302
![]() |
Novus Biologicals
NBP2526430.2MG |
0.2 mg |
Each for $584.50
|
|
|||||
Description
CD3 epsilon Monoclonal specifically detects CD3 epsilon in Mouse samples. It is validated for Flow Cytometry, Functional, CyTOF-ready.Specifications
| CD3 epsilon | |
| Flow Cytometry 1:10 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| CD3E | |
| IgG | |
| Protein A or G purified | |
| The 25-kDa epsilon chain of the T-cell receptor-associated CD3 complex expressed on thymocytes, mature T lymphocytes, and NK-T cells on many mouse stains. |
| 500A2 | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
| 916 | |
| T cell receptor complexes derived from mouse C6VL-BS thymoma cells (C57BL/Ka) . | |
| Primary | |
| Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title