Learn More
CD3 epsilon Antibody (C363.29B (YCD3-1)), FITC, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP126686
Catalog No. NBP126686
Only null left
Add to Cart
Description
Description
CD3 epsilon Monoclonal specifically detects CD3 epsilon in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay.
Specifications
Specifications
CD3 epsilon
Monoclonal
0.5mg/mL
Flow Cytometry <= 3 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay
CD3E
IL-4 producing Th2 cell lines including D10
0.25 mg
Adaptive Immunity, Apoptosis, Diabetes Research, Immunology, Innate Immunity, Signal Transduction
916
Mouse
IgG2b κ
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
C363.29B (YCD3-1)
FITC
CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Rat
Affinity Purified
RUO
Primary
Mouse CD3 epsilon (Mr 25 kD)
Store at 4C. Do not freeze.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only