Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 epsilon Antibody (PC3/188A) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$658.00
Specifications
Antigen | CD3 epsilon |
---|---|
Clone | PC3/188A |
Concentration | 1 mg/mL |
Dilution | Western Blot 1 - 2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254405100
![]() |
Novus Biologicals
NBP254405100UG |
100 μg |
Each for $658.00
|
|
|||||
NB254405200
![]() |
Novus Biologicals
NBP254405200UG |
200 μg | N/A | N/A | N/A | ||||
Description
CD3 epsilon Monoclonal specifically detects CD3 epsilon in Human, Rat, Porcine, Bovine, Chicken, Equine, Guinea Pig, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
CD3 epsilon | |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
PBS with No Preservative | |
916 | |
A synthetic peptide corresponding to amino acids 156-168 of the cytoplasmic domain of human CD3-? chain | |
Primary | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
PC3/188A | |
Western Blot 1 - 2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat, Pig, Bovine, Chicken, Equine, Rabbit | |
CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
CD3E | |
IgG1 | |
Protein A purified | |
20 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title