Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 epsilon Antibody (TR66), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP197446AF350
Description
CD3 epsilon Monoclonal specifically detects CD3 epsilon in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
CD3 epsilon | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
CD3E | |
Cloned human helper T cell line. | |
Primary | |
Recognizes the epsilon-chain of human CD3. | |
Store at 4°C in the dark. |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
TR66 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
Mouse | |
0.1 mL | |
916 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction