Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                CD3 epsilon Antibody (TR66) - BSA Free, Novus Biologicals™
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Mouse Monoclonal Antibody has been used in 5 publications
$206.50 - $532.07
Specifications
| Antigen | CD3 epsilon | 
|---|---|
| Clone | TR66 | 
| Concentration | 1.0 mg/mL | 
| Dilution | Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500 | 
| Applications | Immunofluorescence, Flow Cytometry, Immunocytochemistry, Immunoprecipitation | 
Description
CD3 epsilon Monoclonal specifically detects CD3 epsilon in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| CD3 epsilon | |
| 1.0 mg/mL | |
| Immunofluorescence, Flow Cytometry, Immunocytochemistry, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| CD3E | |
| IgG1 | |
| Protein G purified | |
| Recognizes the epsilon-chain of human CD3. | 
| TR66 | |
| Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 916 | |
| Cloned human helper T cell line. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | 
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title