Learn More
CD3 epsilon Antibody (TR66), Janelia Fluor™ 549, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP197446JF549
Catalog No. NBP97446549
Only null left
Add to Cart
Description
Description
CD3 epsilon Monoclonal specifically detects CD3 epsilon in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
CD3epsilon
Monoclonal
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CD3E
Cloned human helper T cell line.
0.1 mL
Primary
Recognizes the epsilon-chain of human CD3.
Store at 4°C in the dark.
IgG1
Flow Cytometry, Immunocytochemistry, Immunofluorescence
TR66
Flow Cytometry, Immunocytochemistry/Immunofluorescence
CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Mouse
Protein G purified
Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Lines, Stem Cell Markers
916
Human
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.