Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 epsilon Rabbit anti-Human, Clone: 410, Novus Biologicals™

Rabbit Monoclonal Antibody
$342.00 - $524.00
Specifications
Antigen | CD3 epsilon |
---|---|
Clone | 410 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
CD3 epsilon Monoclonal specifically detects CD3 epsilon in Human samples. It is validated for ELISA.Specifications
CD3 epsilon | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
Recombinant Human CD3 epsilon Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
410 | |
ELISA | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
0.2 um filtered solution in PBS | |
916 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title