Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 epsilon, Rabbit anti-Human,Mouse,Rat,Porcine,Amphibian,Bovine,Canine,Chicken,Equine,Feline,Guinea Pig,Hamster,Primate,Rabbit,Sheep, DyLight 594, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP233116DL594
Description
CD3 epsilon Polyclonal specifically detects CD3 epsilon in Human, Rat, Primate samples. It is validated for Western Blot.Specifications
CD3 epsilon | |
Polyclonal | |
Western Blot | |
CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Amphibian, Bovine, Canine, Chicken, Equine, Feline, Guinea Pig, Hamster, Primate, Rabbit | |
IgG |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
DyLight 594 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD3E | |
A synthetic peptide aa156-168 (ERPPPVPNPDYEP) from human CD3e | |
0.1 mL | |
Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
Recognizes the epsilon-chain of CD3, which consists of five different polypeptide chains (designated as gamma, delta, epsilon, zeta, and eta) with MW ranging from 16-28kDa. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction