Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD3/HLA Class 2 Antibody Cocktail, FITC, PE
GREENER_CHOICE

Catalog No. PIMA516958
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516958 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516958 Supplier Invitrogen™ Supplier No. MA516958
Add to Cart
Add to Cart

Antibody Cocktail

MA5-16958 contains a monoclonal mixture of FITC-conjugated CD3 (clone #UCH-T1, Mouse IgG1) and RPE-conjugated HLA Class 2 (clone #WR18, Mouse IgG2a). UCHT1 recognizes the human CD3 cell surface antigen expressed by T lymphocytes. WR18 recognizes a common determinant on the beta chain of the human HLA Class 2 molecules HLA-DR, DP and DQ. This antigen is expressed on B lymphocytes, monocytes and on activated T lymphocytes. This antibody is designed to permit the simultaneous enumeration of CD3 and HLA Class 2 expressing cells. It is particularly useful in the assessment of HLA Class 2 expression on T lymphocytes. Reconstitute with 0.5 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.

The protein encoded by this gene is part of the T-cell receptor/CD3 complex (TCR/CD3 complex) and is involved in T-cell development and signal transduction. The encoded membrane protein represents the delta subunit of the CD3 complex, and along with four other CD3 subunits, binds either TCR alpha/beta or TCR gamma/delta to form the TCR/CD3 complex on the surface of T-cells. Defects in this gene are a cause of severe combined immunodeficiency autosomal recessive T-cell-negative/B-cell-positive/NK-cell-positive (SCIDBNK). Two transcript variants encoding different isoforms have been found for this gene. Other variants may also exist, but the full-length natures of their transcripts has yet to be defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3/HLA Class 2
Applications Flow Cytometry
Classification Cocktail
Clone UCHT1/WR18
Concentration Conc. Not Determined
Conjugate FITC/PE
Formulation PBS with 1% BSA, 5% sucrose and 0.09% sodium azide
Gene CD3G
Gene Accession No. P01911, P01920, P04234, P04440, P07766, P09693
Gene Alias beta1 domain MHC class II HLA DPB; CD3 antigen, delta subunit; CD3 delta; CD3 epsilon; CD3D; CD3d antigen, delta polypeptide (TiT3 complex); CD3d molecule; CD3d molecule, delta (CD3-TCR complex); CD3-DELTA; CD3E; CD3e antigen, epsilon polypeptide (TiT3 complex); CD3e molecule; CD3e molecule, epsilon (CD3-TCR complex); CD3-epsilon; CD3G; CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule; CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); CD3-GAMMA; CD74 antigen; CELIAC1; class II HLA beta chain; Clone P2-beta-3; DHLAG; DPB1; DR-16; DRB1; DRw10; DW2.2/DR2.2; FLJ18683; HLA class II antigen; HLA class II histocompatibility antigen, DP beta 1 chain; HLA class II histocompatibility antigen, DP(W4) beta chain; HLA class II histocompatibility antigen, DQ beta 1 chain; HLA class II histocompatibility antigen, DR-1 beta chain; HLA class II histocompatibility antigen, DRB1-15 beta chain; HLA class II histocompatibility antigen, DRB1-16 beta chain; HLA class II histocompatibility antigen, DRB1-3 chain; HLA DP14-beta chain; HLADG; HLA-DP; HLA-DP histocompatibility type, beta-1 subunit; HLA-DP1B; HLA-DPB; HLA-DPB1; HLA-DQ; HLA-DQB; HLA-DQB1; HLA-DR1B; HLA-DRB; HLA-DRB1; HLA-DRB2; HLA-DR-gamma; human leucocyte antigen DRB1; IA-gamma; IDDM1; IMD17; IMD18; IMD19; lymphocyte antigen; lymphocyte antigen DRB1; major histocompatibility complex class II antigen beta chain; major histocompatibility complex, class II, DP beta 1; major histocompatibility complex, class II, DQ beta 1; major histocompatibility complex, class II, DR beta 1; MHC class II antigen; MHC class II antigen beta chain; MHC class II antigen DP beta 1 chain; MHC class II antigen DPB1; MHC class II antigen DPbeta1; MHC class II antigen DQB1; MHC class II antigen DRB1*15; MHC class II antigen DRB1*16; MHC class II antigen DRB1*3; MHC class II antigen HLA-DQ-beta-1; MHC class II DQ beta chain; MHC class II HLA-DP-beta-1; MHC class II HLA-DQ beta glycoprotein; MHC class II HLA-DR beta 1 chain; MHC class II HLA-DRB1; MHC class II HLA-DR-beta cell surface glycoprotein; MHC class II HLA-DRw10-beta; MHC class2 antigen; MHC DQ beta; MHC HLA DPB1; OKT3, delta chain; SS1; T3D; T3E; T3G; T-cell antigen receptor complex, epsilon subunit of T3; T-cell antigen receptor complex, gamma subunit of T3; T-cell receptor T3 delta chain; T-cell receptor T3 gamma chain; T-cell surface antigen T3/Leu-4 epsilon chain; T-cell surface glycoprotein CD3 delta chain; T-cell surface glycoprotein CD3 epsilon chain; T-cell surface glycoprotein CD3 gamma chain; TCRE
Gene Symbols CD3D, CD3E, CD3G, HLA-DPB1, HLA-DQB1, HLA-DRB1
Host Species Mouse
Immunogen Human infant thymocytes and lymphocytes from a patient with Sezary Syndrome, Human B-CLL cells.
Purification Method Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3115, 3119, 3123, 915, 916, 917
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Lyophilized
Isotype IgG1, IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.