Learn More
Human CD3 (Research Grade Teplizumab Biosimilar) Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Human Monoclonal Antibody
Supplier:  R&D Systems FAB9929S100UG
Catalog No. FAB9929S100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
CD3 Monoclonal specifically detects CD3 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD3
Monoclonal
Alexa Fluor 750
Supplied 0.2 mg/ml in a saline solution containing BSA and Sodium Azide.
Human
Protein A or G purified from cell culture supernatant
Primary
Detects Human CD3 based on Teplizumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Teplizumab. This product is for research use only.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Flow Cytometry
Hu113
Flow Cytometry 0.25-1 μg/10^6 cells
CD3 antigen, delta subunit, CD3d antigen, CD3d antigen, delta polypeptide (TiT3 complex), CD3d molecule, delta (CD3-TCR complex), CD3-DELTA, CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, CD3G, CD3g antigen, CD3g antigen, gamma polypeptide (TiT3 complex), CD3g molecule, epsilon (CD3-TCR complex), CD3g molecule, gamma (CD3-TCR complex), CD3-GAMMA, FLJ17620, FLJ17664, FLJ18683, FLJ79544, FLJ94613, MGC138597, T3DOKT3, delta chain, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell receptor T3 delta chain, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 delta chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Human CD3
100 μg
916
Human
Supernatant
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.