Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 Molecular Complex Rat anti-Mouse, R718, Clone: 17A2, BD Horizon™

Catalog No. BDB567296 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB567296 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB567296 Supplier BD Biosciences Supplier No. 567296
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD3 Molecular Complex Monoclonal antibody specifically detects CD3 Molecular Complex in Mouse samples. It is validated for Flow cytometry (Routinely Tested)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3 Molecular Complex
Applications Flow Cytometry
Classification Monoclonal
Clone 17A2
Concentration 0.2 mg/mL
Conjugate R718
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD3; CD3 epsilon; Cd3e; CD3ε; T3e
Host Species Rat
Immunogen γδ TCR-positive T-T hybridoma D1
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12501
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.