Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 Mouse anti-Baboon,Cynomolgus,Rhesus Ma,Human, BB700, Clone: SP34-2, BD Horizon™

Catalog No. BDB566518 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566518 25 μg
BDB566517 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB566518 Supplier BD Biosciences Supplier No. 566518
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD3 Monoclonal antibody specifically detects CD3 in Baboon,Cynomolgus,Rhesus Ma,Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3
Applications Flow Cytometry
Classification Monoclonal
Clone SP34-2
Concentration 0.2 mg/mL
Conjugate Brilliant Blue 700 (BB700)
Host Species Mouse
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Rhesus Macaque, Human
Content And Storage Refrigerate (2 to 8°C)
Form Purified
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.