Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 Mouse anti-Chicken, Pigeon, Pacific Blue, Clone: CT-3, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 820026
Description
SouthernBiotech is a leading provider of anti-chicken primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
| CD3 | |
| Monoclonal | |
| 0.5mg/mL | |
| Supplied in PBS/NaN3 | |
| Chicken thymocytes and Ig- blood mononuclear cells | |
| RUO | |
| Chicken, Pigeon | |
| IgG1 |
| Flow Cytometry | |
| CT-3 | |
| Pacific Blue | |
| Mouse | |
| 0.1 mg | |
| Primary | |
| Store at 2-8°C; Protect from light |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction