Learn More
CD3 Mouse anti-Human, Alexa Fluor 700, Clone: SP34-2, BD
Mouse Monoclonal Antibody
$138.00 - $307.50
Specifications
Antigen | CD3 |
---|---|
Clone | SP34-2 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Clone SP34-2 is a mouse IgG1 isotype monoclonal antibody, descendant of SP34 (mouse IgG3), with the same specificity and reactivity pattern as the parent clone. It cross-reacts with a major subset of peripheral blood lymphocytes, but not monocytes or granulocytes, of baboon, and rhesus, cynomolgus, and pigtail macaque monkeys. The distribution on lymphocytes is similar to that observed with normal human donor lymphocytes with the majority of CD3-positive cells being negative when dual stained with antibodies to B or NK cells markers. SP34-2 is also capable of inducing cell proliferation on both human and non-human primate PBMC.
Specifications
CD3 | |
0.2mg/mL | |
Monoclonal | |
Mouse | |
Baboon, Cynomolgus, Rhesus Monkey | |
Purified Human CD3ε Protein | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
SP34-2 | |
Flow Cytometry | |
Alexa Fluor 700 | |
RUO | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
IgG1 λ | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.