Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 Mouse anti-Human, Brilliant Ultraviolet 496, Clone: UCHT1, BD Horizon™
Mouse Monoclonal Antibody
$248.00 - $470.00
Specifications
Antigen | CD3 |
---|---|
Clone | UCHT1 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 496 |
Description
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow cytometry.Specifications
CD3 | |
Flow Cytometry | |
Brilliant Ultraviolet 496 | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human infant thymocytes and peripheral blood lymphocytes from a Sézary Syndrome donor | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
UCHT1 | |
Monoclonal | |
Mouse | |
Human | |
CD3e; CD3E; T3E; TCRE; T-cell surface antigen T3/Leu-4 epsilon | |
IgG1 κ | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title