Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 Mouse anti-Human, BUV805, Clone: SK7, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 612893
Description
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow Cytometry.
Specifications
CD3 | |
Monoclonal | |
Brilliant Ultraviolet 805 | |
100 Tests | |
Primary | |
Human | |
IgG1 κ |
Flow Cytometry | |
SK7 (also known as Leu-4) | |
Mouse | |
RUO | |
CD3 | |
Refrigerate (2 to 8°C) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction