Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 Mouse anti-Human, BUV805, Clone: SK7, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 612894
Description
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD3 | |
| Monoclonal | |
| Brilliant Ultraviolet 805 | |
| 25 Tests | |
| Primary | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| SK7 (also known as Leu-4) | |
| Mouse | |
| RUO | |
| CD3 | |
| Refrigerate (2 to 8°C) |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction