Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 Mouse anti-Human, BUV805, Clone: UCHT1, BD Horizon™
Mouse Monoclonal Antibody
$233.00 - $470.00
Specifications
Antigen | CD3 |
---|---|
Clone | UCHT1 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Brilliant Ultraviolet 805 |
Description
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow Cytometry.Specifications
CD3 | |
Flow Cytometry | |
Brilliant Ultraviolet 805 | |
RUO | |
IgG1 κ | |
Refrigerate (2 to 8°C) |
UCHT1 | |
Monoclonal | |
Mouse | |
Human | |
Primary | |
CD3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title