Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 Mouse anti-Human, Clone: OKT3, BD Pharmingen™
Mouse Monoclonal Antibody
$158.70 - $777.50
Specifications
Antigen | CD3 |
---|---|
Clone | OKT3 |
Concentration | 1.0 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow Cytometry, ActivationSpecifications
CD3 | |
1.0 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
No azide/low endotoxin: Aqueous buffered solution containing no preservative, sterile filtered(0.2μm pore size membrane). Endotoxin level is ≤0.1 EU/μg (≤0.01 ng/μg) of protein as determined by the LAL assay. | |
Sheep Erythrocyte Rosette-purified Human T Cells | |
Primary |
OKT3 | |
Flow Cytometry | |
Unconjugated | |
Mouse | |
Human | |
CD3E; CD3e; T-cell surface antigen T3/Leu-4 epsilon; T3E; TCRE | |
IgG2a κ | |
Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. This preparation contains no preservatives, thus it should be handled under aseptic conditions. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title