Learn More
CD3 Mouse anti-Human, FITC, Clone: HIT3A, BD
Mouse Monoclonal Antibody
$62.00 - $122.00
Specifications
Antigen | CD3 |
---|---|
Clone | HIT3A |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB561802
|
BD Biosciences
561802 |
25 Tests |
Each for $62.00
|
|
|||||
BDB555339
|
BD Biosciences
555339 |
100 Tests |
Each for $122.00
|
|
|||||
Description
The HIT3a monoclonal antibody specifically binds to the human CD3 ε-chain, a 20 kDa subunit of the CD3/T cell antigen receptor complex found on 70-80% of normal human peripheral blood lymphocytes and 60-85% of thymocytes. Studies from the HLDA Workshop show that this antibody can be mitogenic for T lymphocytes. The CD3 complex plays a role in signal transduction during antigen recognition by the T cell receptor for antigen. HIT3a antibody does not stain intracellular CD3 unlike the other CD3-specific clone, UCHT1.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
CD3 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
HIT3A | |
Monoclonal | |
Mouse | |
Human | |
IgG2a κ | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.