Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 Mouse anti-Human, FITC, Clone: UCHT1, Southern Biotech™

Mouse Monoclonal Antibody
$65.80 - $147.40
Specifications
Antigen | CD3 |
---|---|
Clone | UCHT1 |
Concentration | Lot specific |
Applications | Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Description
SouthernBiotech anti-human primary antibodies are available in a variety of conjugated formats including Alexa Fluor and Pacific Blue.
Specifications
CD3 | |
Lot specific | |
Monoclonal | |
Mouse | |
Human | |
Human peripheral blood lymphocytes from a patient with Sézary's syndrome and infant thymocytes | |
Primary |
UCHT1 | |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
FITC | |
RUO | |
Supplied in PBS/NaN3 | |
IgG1 | |
Store at 2-8°C; Protect from light |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title