Learn More
CD3 Mouse anti-Rat, FITC, Clone: 1F4, BD
Mouse Monoclonal Antibody
$100.00 - $361.50
Specifications
Antigen | CD3 |
---|---|
Clone | 1F4 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB561801
|
BD Biosciences
561801 |
50 μg |
Each for $100.00
|
|
|||||
BDB557354
|
BD Biosciences
557354 |
0.5 mg |
Each for $361.50
|
|
|||||
Description
The 1F4 antibody reacts with the T-cell receptor-associated CD3 cell-surface antigen found on thymocytes, peripheral T lymphocytes, and dendritic epidermal T cells. In vitro, immobilized 1F4 mAb induces T-cell proliferation, and soluble mAb synergizes with PMA for T-cell activation. Pre-incubation of splenocytes with mAb 1F4 blocks staining with mAb G4.18 (Cat. No. 559974/554830).
This antibody is routinely tested by flow cytometric analysis. Other applications were routinely tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
CD3 | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
Rat | |
F344 rat splenic T cells stimulated with PMA plus A23187 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
1F4 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgM κ | |
Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.