Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 Mouse anti-Rat, FITC, Clone: G4.18, BD

Catalog No. BDB554832 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554832 0.5 mg
BDB559975 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB554832 Supplier BD Biosciences Supplier No. 554832
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The G4.18 antibody reacts with the T-cell receptor-associated CD3 cell-surface antigen found on thymocytes, peripheral T lymphocytes, and dendritic epidermal T cells. It has been reported that CD3 expression is down-regulated within 24 hours in concanavalin A-stimulated rat T cells, and soluble mAb inhibits the allogeneic mixed-lymphocyte proliferative response and cell-mediated cytotoxicity to allogeneic target cells. In vivo treatment with G4.18 mAb prevents cardiac and skin allograft rejection, resulting in donor-specific tolerance. Pre-incubation of splenocytes with the alternate anti-rat CD3 monoclonal antibody, 1F4 (Cat. No. 556970), blocks staining with mAb G4.18.

This antibody is routinely tested by flow cyometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3
Applications Flow Cytometry
Classification Monoclonal
Clone G4.18
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Mouse
Immunogen PHA-stimulated rat lymph node and spleen cells
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG3 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.