Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 Rat anti-Mouse, BV480, Clone: 17A2, BD Horizon™

Catalog No. BDB565642 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565642 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565642 Supplier BD Biosciences Supplier No. 565642
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD3 Monoclonal antibody specifically detects CD3 in Mouse samples. It is validated for Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD3
Applications Immunofluorescence
Classification Monoclonal
Clone 17A2
Concentration 0.2mg/mL
Conjugate Brilliant Violet 480
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias CD3; CD3 epsilon; Cd3e; CD3ε; T3e
Host Species Rat
Immunogen γδ TCR-positive T-T hybridoma D1
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV480 under optimum conditions and unconjugated antibody and free BD Horizon BV480 were removed.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Reseach Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.