Learn More
CD3 zeta Antibody (H146-968), PE, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP144683PE
Description
CD3 zeta Monoclonal specifically detects CD3 zeta in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
CD3 ζ | |
Monoclonal | |
PE | |
Synthetic peptide corresponding to amino acids 151-164 of mouse CD3 zeta. | |
RUO | |
919 | |
Human, Mouse | |
IgG2 κ |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
H146-968 | |
Armenian Hamster | |
0.1 mL | |
Primary | |
The hamster antibody H146-968 reacts with CD3 zeta chain (CD247), which is a component of TCR/CD3 complex expressed on T cells. | |