Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD3 zeta Antibody, Novus Biologicals™
SDP

Catalog No. p-200045605 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB432256 25 μL
NBP185750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB432256 Supplier Novus Biologicals Supplier No. NBP18575025UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

CD3 zeta Polyclonal specifically detects CD3 zeta in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CD3 zeta
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias CD247 antigen, CD247 antigen, zeta subunit, CD247 molecule, CD3H, CD3Q, CD3z antigen, zeta polypeptide (TiT3 complex), CD3ZT-cell receptor T3 zeta chain, T3Z, T-cell antigen receptor complex, zeta subunit of CD3, T-cell surface glycoprotein CD3 zeta chain, TCRZCD3-ZETA
Gene Symbols CD247
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:KFSRSADAPAYQQGQNQLYNELNLGRREEYDVLDKRRGRDPEMGGKPRRKNPQEGLYNELQKDKMAEAYSEIGMKGERRRGKGHDGLYQGLSTATKDTYDALHMQALP
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Innate Immunity, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 919
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.