Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD3 zeta, p Tyr111 Antibody (EM-55), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237709AF350
Description
CD3 zeta Monoclonal specifically detects CD3 zeta in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Flow (Intracellular).Specifications
CD3 zeta | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
CD247 | |
BP1 (pTyr111/123) - KLH. | |
Primary | |
Antibody recognizes phosphorylated tyrosine 111 of CD3 zeta chain (CD247), which is a component of TCR/CD3 complex expressed on T cells. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
EM-55 | |
Western Blot, Flow Cytometry, Flow (Intracellular) | |
CD247 antigen, CD247 antigen, zeta subunit, CD247 molecule, CD3H, CD3Q, CD3z antigen, zeta polypeptide (TiT3 complex), CD3ZT-cell receptor T3 zeta chain, T3Z, T-cell antigen receptor complex, zeta subunit of CD3, T-cell surface glycoprotein CD3 zeta chain, TCRZCD3-ZETA | |
Mouse | |
0.1 mL | |
919 | |
Human, Mouse | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction