Learn More
Invitrogen™ CD30 Monoclonal Antibody (Ber-H2)

Description
MA1-26282 detects CD30 from human samples.
Specifications
Specifications
| Antigen | CD30 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | Ber-H2 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Tissue culture supernatant with 0.05% sodium azide |
| Gene | TNFRSF8 |
| Gene Accession No. | P28908 |
| Gene Alias | Ber-H2; Cd30; CD30 antigen; CD30 molecule; CD30L receptor; cytokine receptor CD30; D1S166E; Ki; Ki-1; Ki-1 antigen; Lymphocyte activation antigen CD30; sCD30; soluble CD30; TNF receptor superfamily member 8; TNFRSF8; Tumor necrosis factor receptor superfamily member 8; tumor necrosis factor receptor superfamily, member 8; Tumor necrosis factor superfamily member 8; tumor necrosis factor superfamily, member CD30 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.