Learn More
CD30 Mouse anti-Human, Brilliant Violet 605, Clone: BerH8 (also known as Ber-H8), BD Optibuild™
Shop All BD Biosciences ProductsDescription
The BerH8 monoclonal antibody specifically binds to CD30, a 120 kDa type I transmembrane glycoprotein expressed on stimulated T and B cells. CD30 is an activation marker initially identified to be expressed on Reed-Sternberg cells. It is also expressed on a few extrafollicular T and B cells located at the rim of germinal centers. CD30 serves as a cytokine receptor. It belongs to the nerve growth factor receptor/tumor necrosis factor receptor (NGFR/TNFR) superfamily and is also known as TNFRSF8. CD30 interaction with CD30 ligand (CD30L/CD153/TNFSF8) can mediate signals for proliferation, apoptosis and cytotoxicity of lymphoid cells.
Specifications
Specifications
| Antigen | CD30 |
| Classification | Monoclonal |
| Clone | BerH8 (also known as Ber-H8) |
| Concentration | 0.2mg/mL |
| Conjugate | Brilliant Violet 605 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | TNFRSF8, TNR8, CD30L/CD153 receptor, Ki-1, D1S166E |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.